2,4,6-Trichloropyrimidine 98%

Supplier: Ambeed
Warning

A133539-100G A133539-10G A133539-500G A133539-5G A133539-25G
77272-110EA 47.38 USD
77272-110 77272-112 77272-114 77298-636 77314-548
2,4,6-Trichloropyrimidine 98%
2,4,6-Trichloropyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₃N₂
MW: 183.42 g/mol
Boiling Pt: 212.7 °C (760 mm Hg)
Melting Pt: 23…25 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00006063
CAS Number: 3764-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR