2,4,6-Trichloropyridine 98%

Supplier: Ambeed
Warning

A133139-100G A133139-10G A133139-1G A133139-25G A133139-5G A133139-500G
103801-834EA 7 USD
103801-834 77234-588 77234-590 77314-448 77314-450 77314-452 77745-953
2,4,6-Trichloropyridine 98%
2,4,6-Trichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₃N
MW: 182.44 g/mol
Melting Pt: 33…37 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00971928
CAS Number: 16063-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR