2,4,6-Trichloropyridin-3-amine 96%

Supplier: Ambeed
Warning

A289171-250MG A289171-5G
103813-816EA 147 USD
103813-816 76827-676 77683-863
2,4,6-Trichloropyridin-3-amine 96%
2,4,6-Trichloropyridin-3-amine

Precaución: For research use only.

Formula: C₅H₃Cl₃N₂
MW: 197.45 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20703662
CAS Number: 91872-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR