2,4,6-Trichloronicotinaldehyde 97%

Supplier: Ambeed
Warning

A132374-100MG A132374-250MG
103801-730EA 108 USD
103801-730 76983-790 76983-792
2,4,6-Trichloronicotinaldehyde 97%
2,4,6-Trichloronicotinaldehyde

Precaución: for Research Use Only

Formula: C₆H₂Cl₃NO
MW: 210.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17012148
CAS Number: 1261269-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR