2,4,6-Trichlorobenzaldehyde 97%

Supplier: Ambeed
Warning

A171276-10G A171276-250MG A171276-25G A171276-1G A171276-5G
103806-616EA 8 USD
103806-616 103844-890 76974-016 76974-018 76974-020 77082-030 77082-032
2,4,6-Trichlorobenzaldehyde 97%
2,4,6-Trichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₃O
Storage Temperature: Refrigerator
MDL Number: MFCD12024961
CAS Number: 24473-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR