2,4,6-Trichloro-5-methylpyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-2,4,6-trichloropyrimidine

A170992-10G A170992-250MG A170992-1G A170992-25G A170992-5G
103806-572EA 10 USD
103806-572 103844-846 76985-414 76985-416 77081-972 77081-974 77081-976
2,4,6-Trichloro-5-methylpyrimidine 98%
2,4,6-Trichloro-5-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₃Cl₃N₂
MW: 197.45 g/mol
Melting Pt: 65…69 °C
Flash Pt: 117.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00233525
CAS Number: 1780-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR