2,4,6-Trichloro-5-methoxypyrimidine 95%

Supplier: Ambeed
Warning

A266399-100MG A266399-250MG A266399-1G A266399-25G
103812-800EA 127 USD
103812-800 103852-702 76988-248 76988-250 77304-954 77683-816
2,4,6-Trichloro-5-methoxypyrimidine 95%
2,4,6-Trichloro-5-methoxypyrimidine

Precaución: For research use only.

Formula: C₅H₃Cl₃N₂O
MW: 213.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11226168
CAS Number: 60703-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR