2,4,6-Trichloro-3-nitropyridine 97%

Supplier: Ambeed
Warning

A112334-10G A112334-250MG A112334-25G A112334-1G A112334-5G
103797-998EA 43 USD
103797-998 103823-832 76982-790 76982-792 76982-794 77284-224 77284-226
2,4,6-Trichloro-3-nitropyridine 97%
2,4,6-Trichloro-3-nitropyridine

Precaución: for Research Use Only

Formula: C₅HCl₃N₂O₂
MW: 227.433 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09264535
CAS Number: 60186-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR