2,4,6-Tribromophenyl acrylate 98%

Supplier: Ambeed
Warning

Synonyms: Acrylic Acid 2,4,6-Tribromophenyl Ester

A180985-5G A180985-1G
103807-578EA 7 USD
103807-578 103845-940 77083-456 77301-702
2,4,6-Tribromophenyl acrylate 98%
2,4,6-Tribromophenyl acrylate

Precaución: for Research Use Only

Formula: C₉H₅Br₃O₂
MW: 384.85 g/mol
Boiling Pt: 380.8 °C (760 mm Hg)
Flash Pt: 184.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00078430
CAS Number: 3741-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR