2,4,5-Trimethoxybenzaldehyde 98%

Supplier: Ambeed
Warning

A123250-100G A123250-100MG A123250-10G A123250-25G A123250-500G
77094-564EA 45.62 USD
77094-564 77094-566 77094-568 77094-570 77094-572
2,4,5-Trimethoxybenzaldehyde 98%
2,4,5-Trimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 334.7 °C (760 mm Hg)
Melting Pt: 111…115 °C
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00003312
CAS Number: 4460-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR