2,4,5-Trichloropyrimidine 95%

Supplier: Ambeed
Warning

A171599-100G A171599-10G A171599-1G A171599-500G A171599-25G A171599-5G
103844-924EA 6 USD
103844-924 76824-524 76824-526 76824-528 76824-530 77082-086 77301-140
2,4,5-Trichloropyrimidine 95%
2,4,5-Trichloropyrimidine

Precaución: for Research Use Only

Formula: C₄HCl₃N₂
MW: 183.42 g/mol
Boiling Pt: 258.5 °C (760 mm Hg)
Flash Pt: 109.9 °C
Storage Temperature: Freezer
MDL Number: MFCD03788200
CAS Number: 5750-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR