2,3-Dioxoindoline-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A178661-100MG A178661-250MG
103807-362EA 214 USD
103807-362 77011-884 77011-886
2,3-Dioxoindoline-4-carboxylic acid 95%
2,3-Dioxoindoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD16876980
CAS Number: 41704-95-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR