2,3-Dimethylhydroquinone 98%
Supplier: Ambeed
|
Warning
|
A122728-10G
A122728-25G
A122728-100G
103800-262EA
22
USD
103800-262
77108-660
77566-332
77571-378
2,3-Dimethylhydroquinone 98%
2,3-Dimethylhydroquinone
Precaución: For research use only.
Formula:
C₈H₁₀O₂ Boiling Pt: 291.5 °C (760 mm Hg) Melting Pt: 223…225 °C Flash Pt: 145.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009997 CAS Number: 608-43-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...