2,3-Dimethylbenzoic acid 97%

Supplier: Ambeed
Warning

A101265-10G A101265-1G A101265-25G A101265-5G
103821-564EA 8 USD
103821-564 77245-104 77245-106 77245-108 77297-032
2,3-Dimethylbenzoic acid 97%
2,3-Dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 281.5 °C (760 mm Hg)
Melting Pt: 144…146 °C
Flash Pt: 129.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002479
CAS Number: 603-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR