2,3-Dimethylphenylacetic acid 95%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dimethylbenzeneacetic acid

A273748-250MG A273748-1G A273748-25G A273748-5G
103813-156EA 30 USD
103813-156 103853-080 76692-898 77092-884 77092-886 77305-146
2,3-Dimethylphenylacetic acid 95%
2,3-Dimethylphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.204 g/mol
Boiling Pt: 300 °C (760 mm Hg)
Melting Pt: 116…117 °C
Flash Pt: 197.2 °C
Storage Temperature: Ambient
MDL Number: MFCD03844736
CAS Number: 30981-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR