2,3-Dimethoxypyridine 97%

Supplier: Ambeed
Warning

A294014-1G A294014-100G A294014-10G A294014-250MG A294014-500G A294014-25G A294014-5G
103813-994EA 7 USD
103813-994 103835-134 76837-754 76977-620 76977-622 76977-624 76977-626 77305-644 77305-646
2,3-Dimethoxypyridine 97%
2,3-Dimethoxypyridine

Precaución: For research use only

Formula: C₇H₉NO₂
Boiling Pt: 179.8 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD03095043
CAS Number: 52605-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR