2,3-Dimethoxy-5-methyl-p-benzoquinone 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione, 2,3-Dimethoxy-5-methyl-1,4-benzoquinone

A133801-100G A133801-10G A133801-25G A133801-5G
76811-374EA 54.11 USD
76811-374 76811-376 77314-604 77314-606
2,3-Dimethoxy-5-methyl-p-benzoquinone 98%
2,3-Dimethoxy-5-methyl-p-benzoquinone

Precaución: for Research Use Only

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 58…60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001595
CAS Number: 605-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR