2,3-Dihydroxypropyl benzoate 95%
Supplier: Ambeed
|
Warning
|
A214057-100MG
A214057-250MG
A214057-1G
103818-362EA
135
USD
103818-362
103849-070
77165-046
77165-048
77303-322
2,3-Dihydroxypropyl benzoate 95%
2,3-Dihydroxypropyl benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂O₄ Boiling Pt: 376.6 °C (760 mm Hg) Flash Pt: 151.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD01084321 CAS Number: 3376-59-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...