2,3-Dihydroxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: Naphthalene-2,3-diol, 2,3-Naphthalenediol

A614711-25G A614711-100G A614711-500G A614711-5G
103840-186EA 8 USD
103840-186 76844-348 77144-982 77144-984 77307-950
2,3-Dihydroxynaphthalene 98%
2,3-Dihydroxynaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₈O₂
MW: 160.17 g/mol
Boiling Pt: 353.9 °C (760 mm Hg)
Melting Pt: 162…166 °C (760 mm Hg)
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00004073
CAS Number: 92-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR