3,4-Ethylenedioxythiophene 98%
Supplier: Ambeed
|
Danger
|
A408207-25G
A408207-5G
A408207-100G
A408207-10G
103838-222EA
5
USD
103838-222
76841-726
76841-728
77115-788
77115-790
3,4-Ethylenedioxythiophene 98%
3,4-Ethylenedioxythiophene
Precaución: for Research Use Only
Formula:
C₆H₆O₂S Boiling Pt: 193 °C (760 mm Hg) Melting Pt: 10.5 °C Flash Pt: 104 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02093622 CAS Number: 126213-50-1 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...