2,3-Dihydroquinolin-4(1H)-one hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 1,2,3,4-Tetrahydro-4-quinolinone hydrochloride

A112726-250MG A112726-1G A112726-5G
103798-088EA 62 USD
103798-088 103823-926 76810-002 77310-744 77310-746
2,3-Dihydroquinolin-4(1H)-one hydrochloride 95%
2,3-Dihydroquinolin-4(1H)-one hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO
MW: 183.6400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11052533
CAS Number: 71412-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR