2,3-Dihydroquinolin-4(1H)-one 96%

Supplier: Ambeed
Warning

A115583-250MG A115583-5G A115583-1G
103798-590EA 36 USD
103798-590 103824-448 77160-988 77297-380 77311-264
2,3-Dihydroquinolin-4(1H)-one 96%
2,3-Dihydroquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO
MW: 147.17 g/mol
Boiling Pt: 301 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00666394
CAS Number: 4295-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR