2,3-Dihydro-1,4-isoquinolinedione 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydroisoquinoline-1,4-dione

A140596-100MG A140596-250MG
103802-886EA 123 USD
103802-886 77162-290 77162-292
2,3-Dihydro-1,4-isoquinolinedione 97%
2,3-Dihydro-1,4-isoquinolinedione

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13179340
CAS Number: 31053-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR