5-Amino-2,3-dihydrobenzo[b]furan 97%

Supplier: Ambeed
Warning

A198883-100MG A198883-10G A198883-250MG A198883-1G A198883-5G
103817-054EA 16 USD
103817-054 103847-682 76814-190 76814-192 76814-194 77085-706 77085-708
5-Amino-2,3-dihydrobenzo[b]furan 97%
5-Amino-2,3-dihydrobenzo[b]furan

Precaución: for Research Use Only

Formula: C₈H₉NO
Boiling Pt: 289.4 °C (760 mm Hg)
Melting Pt: 78…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03617969
CAS Number: 42933-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR