2,3-Dihydro-1-benzofuran-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A251217-1G A251217-100MG A251217-250MG A251217-10G
103811-972EA 23 USD
103811-972 103851-814 77091-190 77261-992 77261-994 77748-472
2,3-Dihydro-1-benzofuran-2-carboxylic acid 97%
2,3-Dihydro-1-benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
Boiling Pt: 340.5 °C (760 mm Hg)
Melting Pt: 116…118 °C
Flash Pt: 144.317 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03086168
CAS Number: 1914-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR