2,3-Dihydrobenzo[b]furan-5-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 5-Coumarancarboxaldehyde, 2,3-Dihydrobenzofuran-5-carbaldehyde, 2,3-Dihydrobenzo[b]furan-5-carboxaldehyde, 2,3-Dihydrobenzofuran-5-carboxaldehyde

A106859-100G A106859-10G A106859-25G A106859-1G A106859-5G
103797-088EA 6 USD
103797-088 103822-844 77257-470 77257-472 77257-474 77309-700 77309-702
2,3-Dihydrobenzo[b]furan-5-carbaldehyde 97%
2,3-Dihydrobenzo[b]furan-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 277.5 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00068058
CAS Number: 55745-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR