1,4-Benzodioxane-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine-2-carboxylic acid

A176169-100G A176169-10G A176169-25G A176169-5G
103845-438EA 6 USD
103845-438 77191-558 77191-560 77301-432 77301-434
1,4-Benzodioxane-2-carboxylic acid 98%
1,4-Benzodioxane-2-carboxylic acid

Precaución: Sealed in dry, at room temperature.

Formula: C₉H₈O₄
MW: 180.16 g/mol
Boiling Pt: 347.2 °C (760 mm Hg)
Melting Pt: 126…130 °C
Flash Pt: 145.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00084888
CAS Number: 3663-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR