1,4-Benzodioxane 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydrobenzo[b][1,4]dioxine, 2,3-Dihydro-1,4-benzodioxin, Benzo-1,4-dioxane, 1,4-Benzodioxan

A147849-100G A147849-10G A147849-500G A147849-25G A147849-5G
103841-952EA 5 USD
103841-952 77095-724 77095-726 77095-728 77299-604 77299-606
1,4-Benzodioxane 98%
1,4-Benzodioxane

Precaución: for Research Use Only

Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 248.138 °C (760 mm Hg)
Flash Pt: 87.778 °C
Storage Temperature: Ambient
MDL Number: MFCD00006821
CAS Number: 493-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR