2,3-Dihydro-1H-pyrrolo[2,3-c]pyridine 98%
Supplier: Ambeed
|
Warning
|
A193246-100MG
A193246-250MG
A193246-1G
103816-516EA
131
USD
103816-516
103847-130
77029-882
77029-884
77085-026
2,3-Dihydro-1H-pyrrolo[2,3-c]pyridine 98%
2,3-Dihydro-1H-pyrrolo[2,3-c]pyridine
Precaución: for Research Use Only
Formula:
C₇H₈N₂ Boiling Pt: 240.1 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD09907942 CAS Number: 760919-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...