2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dihydro-7-azaindole

A156574-100G A156574-10G A156574-25G A156574-5G A156574-1G
103804-904EA 17 USD
103804-904 103843-104 77247-496 77247-498 77300-198 77300-200 77318-896
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine 98%
2,3-Dihydro-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₈N₂
MW: 120.15 g/mol
Melting Pt: 78…83 °C
Flash Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD06659751
CAS Number: 10592-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR