(±)-1-Aminoindan 98%

Supplier: Ambeed
Warning

A126079-100G A126079-10G A126079-1G A126079-25G A126079-5G
103800-758EA 7 USD
103800-758 77246-330 77246-332 77312-930 77312-932 77312-934
(±)-1-Aminoindan 98%
(±)-1-Aminoindan

Precaución: for Research Use Only

Formula: C₉H₁₁N
Boiling Pt: 225 °C (760 mm Hg)
Melting Pt: 1.5 °C
Flash Pt: 94.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003799
CAS Number: 34698-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR