2,3-Dihydro-1H-benzo[d]imidazo[1,2-a]imidazole 98%

Supplier: Ambeed
Warning

A143226-250MG A143226-1G A143226-5G
103803-252EA 77 USD
103803-252 77235-086 77316-496 77316-498
2,3-Dihydro-1H-benzo[d]imidazo[1,2-a]imidazole 98%
2,3-Dihydro-1H-benzo[d]imidazo[1,2-a]imidazole

Precaución: for Research Use Only

Formula: C₉H₉N₃
MW: 159.19 g/mol
Boiling Pt: 289.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00805823
CAS Number: 24134-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR