2,3-Difluorophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid
A110790-100G
A110790-10G
A110790-1G
A110790-25G
A110790-500G
A110790-5G
103797-734EA
5
USD
103797-734
103823-542
77284-046
77284-048
77284-050
77284-052
77284-054
77310-338
2,3-Difluorophenylboronic acid 97%
2,3-Difluorophenylboronic acid
Precaución: for Research Use Only
Formula:
C₆H₅BF₂O₂ MW: 157.91 g/mol Boiling Pt: 274.8 °C (760 mm Hg) Melting Pt: 95 °C Flash Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD01863170 CAS Number: 121219-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...