1,2-Difluoro-3-nitrobenzene 98%

Supplier: Ambeed
Warning

A199358-1G A199358-25G A199358-5G A199358-100G A199358-10G
103817-100EA 14 USD
103817-100 103847-724 77085-766 77085-768 77085-770 77287-624 77287-626
1,2-Difluoro-3-nitrobenzene 98%
1,2-Difluoro-3-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
Flash Pt: 93.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00456802
CAS Number: 6921-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR