2,3-Difluoro-5-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A278142-25G A278142-5G A278142-1G
103813-396EA 6 USD
103813-396 103853-334 77093-242 77093-244 77305-292
2,3-Difluoro-5-(trifluoromethyl)pyridine 98%
2,3-Difluoro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂F₅N
MW: 183.08 g/mol
Boiling Pt: 104 °C (760 mm Hg)
Flash Pt: 28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972837
CAS Number: 89402-42-6
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR