2,3-Dichloropyrido[2,3-b]pyrazine 95%

Supplier: Ambeed
Warning

A333564-100MG A333564-250MG A333564-1G A333564-5G
103825-650EA 86 USD
103825-650 103836-616 77251-302 77251-304 77306-458 77306-460
2,3-Dichloropyrido[2,3-b]pyrazine 95%
2,3-Dichloropyrido[2,3-b]pyrazine

Precaución: for Research Use Only

Formula: C₇H₃Cl₂N₃
MW: 200.03 g/mol
Boiling Pt: 282.342 °C (760 mm Hg)
Flash Pt: 151.584 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09031107
CAS Number: 25710-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR