2,3-Dichlorophenylboronic acid 98%

Supplier: Ambeed
Warning

A329444-25G A329444-5G A329444-10G A329444-100G
103825-526EA 9 USD
103825-526 103836-480 76839-448 76839-450 77099-968 77685-943
2,3-Dichlorophenylboronic acid 98%
2,3-Dichlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Boiling Pt: 349.8 °C (760 mm Hg)
Melting Pt: 125 °C
Flash Pt: 165.4 °C
Storage Temperature: Freezer
MDL Number: MFCD01075703
CAS Number: 151169-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR