2,3-Dichloroisonicotinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichloropyridine-4-carboxylic acid

A213246-1G A213246-25G A213246-5G A213246-100G A213246-100MG A213246-10G A213246-250MG
103818-278EA 8 USD
103818-278 103848-984 77087-484 77087-486 77087-488 77249-162 77249-164 77249-166 77249-168
2,3-Dichloroisonicotinic acid 98%
2,3-Dichloroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MW: 192 g/mol
Melting Pt: 203…207 °C
Flash Pt: 203.9 °C
Storage Temperature: Ambient
MDL Number: MFCD01861977
CAS Number: 184416-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR