2,3-Dichloro-4-formylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichloropyridine-4-carboxaldehyde, 2,3-Dichloroisonicotinaldehyde

A178457-1G A178457-25G A178457-100MG A178457-250MG
103807-338EA 14 USD
103807-338 103845-676 77083-108 77083-110 77111-320 77111-322
2,3-Dichloro-4-formylpyridine 97%
2,3-Dichloro-4-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176.0000 g/mol
Boiling Pt: 239.7 °C (760 mm Hg)
Melting Pt: 94…96 °C
Flash Pt: 98.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08277282
CAS Number: 884495-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR