2,3-Dichlorofuro[3,4-b]pyrazine-5,7-dione 95%

Supplier: Ambeed
Warning

A142077-100MG A142077-250MG A142077-1G
103803-094EA 160 USD
103803-094 77150-120 77150-122 77316-306
2,3-Dichlorofuro[3,4-b]pyrazine-5,7-dione 95%
2,3-Dichlorofuro[3,4-b]pyrazine-5,7-dione

Precaución: for Research Use Only

Formula: C₆Cl₂N₂O₃
MW: 218.98 g/mol
Boiling Pt: 336.9±37.0 °C (760 mm Hg)
Flash Pt: 157.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD29056530
CAS Number: 144692-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR