2,3-Dichlorobenzoic acid 98%

Supplier: Ambeed
Warning

A129163-100G A129163-500G
77109-110EA 25.47 USD
77109-110 77109-112
2,3-Dichlorobenzoic acid 98%
2,3-Dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 306.9 °C (760 mm Hg)
Melting Pt: 168…170 °C
Flash Pt: 139.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00002413
CAS Number: 50-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR