2,3-Dichlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A744541-1000G A744541-100G A744541-10G A744541-500G A744541-25G
103810-552EA 147 USD
103810-552 103840-830 76845-062 77036-072 77036-074 77036-076 77308-244
2,3-Dichlorobenzaldehyde 98%
2,3-Dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄Cl₂O
Melting Pt: 64…67 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00010127
CAS Number: 6334-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR