2,3-Dichloro-6-methylquinoxaline 98%
Supplier: Ambeed
|
Warning
|
A253263-1G
A253263-5G
A253263-10G
103812-088EA
53
USD
103812-088
103851-936
77091-366
77091-368
77249-982
2,3-Dichloro-6-methylquinoxaline 98%
2,3-Dichloro-6-methylquinoxaline
Precaución: for Research Use Only
Formula:
C₉H₆Cl₂N₂ Boiling Pt: 289.1 °C (760 mm Hg) Melting Pt: 115 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00270819 CAS Number: 39267-05-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...