2,3-Dichloro-6-methoxyquinoxaline 98%

Supplier: Ambeed
Warning

Synonyms: 2,3-Dichloro-6-methoxy-1,4-benzodiazine

A169643-100MG A169643-10G A169643-250MG A169643-5G A169643-1G
103806-392EA 100 USD
103806-392 103844-652 77028-972 77028-974 77028-976 77081-740 77300-982
2,3-Dichloro-6-methoxyquinoxaline 98%
2,3-Dichloro-6-methoxyquinoxaline

Precaución: for Research Use Only

Formula: C₉H₆Cl₂N₂O
MW: 229.063 g/mol
Boiling Pt: 312.6 °C (760 mm Hg)
Flash Pt: 142.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00461063
CAS Number: 39267-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR