2,3-Dichloro-6-methoxypyridine 97%

Supplier: Ambeed
Warning

A198626-10G A198626-250MG A198626-1G A198626-5G
103817-036EA 93 USD
103817-036 103847-664 76814-178 76814-180 77085-686 77085-688
2,3-Dichloro-6-methoxypyridine 97%
2,3-Dichloro-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂NO
MW: 178.02 g/mol
Boiling Pt: 218.6 °C (760 mm Hg)
Melting Pt: 50…54 °C
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669402
CAS Number: 83732-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR