2,3-Dichloro-5-methylpyridine 98%

Supplier: Ambeed
Warning

A166588-100G A166588-250MG A166588-1G A166588-25G A166588-5G
103806-080EA 12 USD
103806-080 103844-344 76813-042 76813-044 77320-608 77320-610 77320-612
2,3-Dichloro-5-methylpyridine 98%
2,3-Dichloro-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N
MW: 162.0166 g/mol
Boiling Pt: 186.9 °C (760 mm Hg)
Melting Pt: 42…45 °C
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD08704667
CAS Number: 59782-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR