2,3-Dichloro-5-formylpyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dichloronicotinaldehyde, 5,6-Dichloropyridine-3-carbaldehyde

A169315-100MG A169315-250MG
103806-370EA 56 USD
103806-370 103844-624 77110-926 77110-928
2,3-Dichloro-5-formylpyridine 95%
2,3-Dichloro-5-formylpyridine

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176.0001 g/mol
Boiling Pt: 268.6 °C (760 mm Hg)
Melting Pt: 69…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08448226
CAS Number: 71690-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR