2,3-Dichloro-5-fluoropyridine 98%

Supplier: Ambeed
Warning

A113700-100G A113700-100MG A113700-10G A113700-250MG A113700-25G A113700-1G A113700-5G
103798-268EA 10 USD
103798-268 103824-110 76781-246 76781-248 76781-250 76781-252 76781-254 77297-258 77297-260
2,3-Dichloro-5-fluoropyridine 98%
2,3-Dichloro-5-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂FN
Boiling Pt: 174 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD08669666
CAS Number: 185985-40-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR