2,3-Dichloro-4-nitroaniline 95%

Supplier: Ambeed
Warning

A817442-100MG A817442-250MG A817442-25G A817442-1G A817442-5G
103841-112EA 33 USD
103841-112 76820-646 76820-648 76820-650 76845-352 76845-354
2,3-Dichloro-4-nitroaniline 95%
2,3-Dichloro-4-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD11226295
CAS Number: 69951-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR