2,3-Dichloro-4-fluoroaniline 95%

Supplier: Ambeed
Warning

A123656-250MG A123656-5G
103800-428EA 22 USD
103800-428 77246-202 77312-544
2,3-Dichloro-4-fluoroaniline 95%
2,3-Dichloro-4-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₄Cl₂FN
MW: 180.0071 g/mol
Boiling Pt: 253.3 °C (760 mm Hg)
Melting Pt: 63…68 °C
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08706409
CAS Number: 36556-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR